Sodium hydrogen carbonate 99+% HPLC

Supplier: Thermo Fisher Scientific

Synonyms: Sodium bicarbonate

446230010 446232500
ACRO446230010EA 113 EUR
ACRO446230010 ACRO446232500
Sodium hydrogen carbonate 99+% HPLC
Sodium hydrogen carbonate
Formula: NaHCO₃
MW: 84.01 g/mol
Melting Pt: 270 °C
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
UV at 500 nm A: ≤0.01
pH 6.5 to 8.5
Titration with HCl ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR