Calcium sulphate dihydrate 98+%, extra_pure

Supplier: Thermo Fisher Scientific

Synonyms: Plaster of Paris

446380010 446380050
ACRO446380010EA 68.2 EUR
ACRO446380010 ACRO446380050
Calcium sulphate dihydrate 98+%, extra_pure
Calcium sulphate dihydrate
Formula: CaSO₄·2H₂O
MW: 172.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149625
CAS Number: 10101-41-4
Merck Index: 13,01711

Order Now

Specification Test Results

Appearance White Powder
Titration Complexometric ≥98.0 %
Water 19 to 23 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR