Sodium acetate, anhydrous 99% for HPLC

Supplier: Thermo Fisher Scientific

Synonyms: Acetic acid sodium salt

446880500 446882500
ACRO446880500EA 53.4 EUR
ACRO446880500 ACRO446882500
Sodium acetate, anhydrous 99% for HPLC
Sodium acetate
Formula: H₃CCOONa
MW: 82.03 g/mol
Boiling Pt: 881.4
Melting Pt: 324 °C
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
Merck Index: 12,08711

Order Now

Specification Test Results

Appearance White Crystalline powder
Loss on drying ≤1 %
UV (0.5 M in water)
Titration with HClO4 ≥98.5 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR