Sodium hydrogen carbonate 99.5% Biochemistry

Supplier: Thermo Fisher Scientific

Synonyms: Sodium bicarbonate

447100010 447102500
ACRO447100010EA 67.5 EUR
ACRO447100010 ACRO447102500
Sodium hydrogen carbonate 99.5% Biochemistry
Sodium hydrogen carbonate
Formula: NaHCO₃
MW: 84.01 g/mol
Melting Pt: 270 °C
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Solubility Clear colorless
UV (10 % in water)
Titration with HCl 99.4 to 100.6 %
Chloride (Cl) ≤20 ppm
Sulfate (SO4) ≤150 ppm
Ammonium (NH4) ≤5 ppm
Calcium (Ca) ≤100 ppm
Copper (Cu) ≤2 ppm
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤50 ppm
Potassium (K) ≤50 ppm
Zinc (Zn) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR