Potassium hydrogen sulphate 99%, extra_pure

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Potassium bisulphate

447610025 447610100 447615000
ACRO447610025EA 190 EUR
ACRO447610025 ACRO447610100 ACRO447615000
Potassium hydrogen sulphate 99%, extra_pure
Potassium hydrogen sulphate
Formula: KHSO₄
MW: 136.17 g/mol
Melting Pt: 214 °C
Storage Temperature: Ambient
MDL Number: MFCD00011404
CAS Number: 7646-93-7
UN: 2509
ADR: 8 II
Merck Index: 13,07695

Order Now

Specification Test Results

Appearance White Crystalline powder
Solubility (10 % in water) Clear colorless
Titration with NaOH ≥98.5 %
Chloride (Cl) ≤0.002 %
Ammonium (NH4) ≤0.003 %
Iron (Fe) ≤0.003 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR