3-Fluoroazetidine hydrochloride 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 3-Fluoroazetidine HCl

449990010
ACRO449990010EA 176 EUR
ACRO449990010
3-Fluoroazetidine hydrochloride 95%
3-Fluoroazetidine hydrochloride
Formula: C3H7ClFN
MW: 111.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663715
CAS Number: 617718-46-4

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Titration Argentometric ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR