Polyvinyl alcohol, M.W. ∼ 85,000-120,000 88%, hydrolysed

Supplier: Thermo Fisher Scientific

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

450140010 450140250 450145000
ACRO450140010EA 283 EUR
ACRO450140010 ACRO450140250 ACRO450145000
Polyvinyl alcohol, M.W. ∼ 85,000-120,000 88%, hydrolysed
Polyvinyl alcohol
Formula: [-CH₂CH(OH)-]n
Melting Pt: 230…240 °C
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Specification Test Results

Appearance White to off-white Granular solid
Infrared spectrum Conforms
Viscosity 23.0 to 27.0 cP (4 % in water)
Degree of hydrolysis 87.0 to 89.0 mol %
Ash ≤0.5 % (Na2O)
pH 4.5 to 6.5 (4 % in water)
Volatile matter ≤5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR