Sodium trifluoromethanesulphinate 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Trifluoromethanesulphinic acid Sodium Salt, Langlois Reagent

454990010 454990050 454990250
ACRO454990010EA 25.9 EUR
ACRO454990010 ACRO454990050 ACRO454990250
Sodium trifluoromethanesulphinate 98%
Sodium trifluoromethanesulphinate
Formula: CHF₃O₂S
MW: 156.05 g/mol
Melting Pt: 350 °C
Storage Temperature: Ambient
MDL Number: MFCD03092989
CAS Number: 2926-29-6

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Water ≤1 %
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR