2'-Deoxycytidine hydrochloride 98%

Supplier: Thermo Fisher Scientific

Synonyms: 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one hydrochloride

458510010 458510050 458510250
ACRO458510010EA 44.4 EUR
ACRO458510010 ACRO458510050 ACRO458510250
2'-Deoxycytidine hydrochloride 98%
2'-Deoxycytidine hydrochloride
Formula: C₉H₁₄ClN₃O₄
MW: 263.68 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00012840
CAS Number: 3992-42-5

Order Now

Specification Test Results

Appearance Off-white to light yellow Crystalline powder
Infrared spectrum Conforms
Water ≤8 %
HPLC ≥97.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR