Lumiracoxib

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-(2-((2-chloro-6-fluorophenyl)amino)-5-methylphenyl)acetic acid, CGS-35189, COX-189,  

462801000 462802500
ACRO462801000EA 126 EUR
ACRO462801000 ACRO462802500
Lumiracoxib
Lumiracoxib

  • Bioactive

Formula: C₁₅H₁₃ClFNO₂
MW: 293.72 g/mol
Melting Pt: 139…141 °C
Storage Temperature: Freezer
MDL Number: MFCD07186254
CAS Number: 220991-20-8

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 %
Sulfated ash ≤0.3 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR