Letrozole

Supplier: Thermo Scientific
Danger

Synonyms: 1-Bis(4-cyanophenyl)methyl-1,2,4-triazole, 4,4'-(1,2,4-Triazol-1-ylmethylene)dibenzonitrile

463190010
ACRO463190010EA 125 EUR
ACRO463190010
Letrozole
Letrozole

  • Bioactive

Formula: C₁₇H₁₁N₅
MW: 285.31 g/mol
Melting Pt: 185 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00866241
CAS Number: 112809-51-5

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 183 to 187 °C
Water ≤0.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR