Aluminium standard solution, 10000 mg/L Al in 5% nitric acid standard for ICP
Supplier: ACTIVE MOTIF
C102.5NP.L1 C102.5NP.L25 C102.5NP.L5
ACSDC102.5NP.L1EA 192 EUR
ACSDC102.5NP.L1 ACSDC102.5NP.L25 ACSDC102.5NP.L5
Aluminium standard solution, 10000 mg/L Al in 5% nitric acid standard for ICP
Standards Cationic Standards Aluminium Standards