Titanium standard solution, 10,000 mg/L Ti in 5% nitric acid with 2% hydrofluoric acid standard for ICP
Supplier: ACTIVE MOTIF
C161.5N2FP.L25 C161.5N2FP.L5
ACSDC161.5N2FP.L25EA 287 EUR
ACSDC161.5N2FP.L25 ACSDC161.5N2FP.L5
Titanium standard solution, 10,000 mg/L Ti in 5% nitric acid with 2% hydrofluoric acid standard for ICP
Standards Cationic Standards Titanium Standards