SCSI supercompetent cells
Supplier: AGILENT
200231
AGLS200231EA 303 EUR
AGLS200231
SCSI supercompetent cells
Competent Cells
SCSI supercompetent and DH5α competent cells are classic E. coli strains engineered for competence.
  • Engineered to become competent
This collection of competent cells, including SCSI supercompetent cells and DH5α competent cells, consists of classic E. coli strains that have been engineered to become competent for use in cloning experiments.