InterPlay adenoviral TAP system
Supplier: AGILENT
240101
AGLS240101EA 789 EUR
AGLS240101
InterPlay adenoviral TAP system
Vectors, Plasmids and Libraries
The InterPlay Mammalian TAP system allows you to recover interacting proteins from mammalian cells.
  • Protease digestion unnecessary to recover intact interacting protein partners since SBP and CBP elute easily from their affinity resins
  • Exceptionally pure protein for downstream analysis
This method is based on expression of a protein of interest fused to two affinity tags: a streptavidin binding peptide (SBP) and a calmodulin binding peptide (CBP). Tandem affinity purification yields your tagged protein and interacting proteins using gentle washing and small molecule elution conditions.
Upozornení: For research use only. Not for use in diagnostic procedures.