Bioanalyzer DNA Analysis
Supplier: AGILENT
5067-1504 5067-1505 5067-1506 5067-1507 5067-1508 5067-1509
AGLS5067-1504EA 1370 EUR
AGLS5067-1504 AGLS5067-1505 AGLS5067-1506 AGLS5067-1507 AGLS5067-1508 AGLS5067-1509
Bioanalyzer DNA Analysis
Electrophoresis Systems Capillary Electrophoresis Systems
DNA fragment or NGS library analysis for sample quality control - for samples up to 12000 bp.
  • High sensitivity from laser (LIF) detection of DNA fragments down to 0,1 ng
  • The broad linear dynamic range allows detection of weak bands without saturation from abundant fragments
  • Sizing and quantitation accuracy and reproducibility results from pre-packaged reagents, standardised assays and automated data analysis
  • Minimal sample consumption is required as only 1 µl is needed for the assay, saving precious material for downstream experiments
  • Fast results are