Capillary Arrays
Supplier: AGILENT
A2300-12502247 A2300-12503355 A2300-12505580 A2300-48503355 A2300-96503355 A2300-96505580
AGLSA2300-12502247EA 1270 EUR
AGLSA2300-12502247 AGLSA2300-12503355 AGLSA2300-12505580 AGLSA2300-48503355 AGLSA2300-96503355 AGLSA2300-96505580
Capillary Arrays
Electrophoresis Systems Capillary Electrophoresis Systems
Capillary arrays for fragment analyser and ZAG systems.
  • Array cartridges are robust and have a long lifespan, keeping your instrument on-task from one project to the next
  • Short arrays provide robust separation for most samples
  • Speed up separation times and increase overall QC throughput with ultra-short arrays
  • Enhance separation resolution and focus nucleic acid analysis with long arrays
The fragment analyser systems offer a selection of capillary arrays to optimize your QC workflow by separating nucleic acids via parallel capillary electrophoresis.More Product Information