ProteoAnalyzer Systems
Supplier: AGILENT
M5350AA M5350-64001
AGLSM5350AAEA 88800 EUR
AGLSM5350AA AGLSM5350-64001
ProteoAnalyzer Systems
Analysers Protein Analysers
Automated SDS capillary electrophoresis system separates 12 protein samples in parallel.
  • Short analysis time - analyse 12 protein samples in parallel in as little as 30 minutes
  • Various sample types - consistently separate challenging protein samples such as membrane proteins and crude lysates
  • High separation resolution - resolve glycosylated and non-glycosylated isoforms broad sizing range - Agilent protein broad range P240 kit for sizing proteins from 10 to 240 kDa
  • High sensitivity - detect as low as 1 ng/µl concentration of BSA protein in PBS
  • Wide sample