Anti-SNRK1 Rabbit Polyclonal Antibody

Supplier: Agrisera
AS214581
AGRIAS214581EA 329 EUR
AGRIAS214581
Anti-SNRK1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: SNRK (SNF related kinase)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Arabidopsis thaliana
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR