Precision thermometer
Supplier: Amarell
M242362-TOL
AMARM242362-TOLEA 58.97 EUR
AMARM242362-TOL
Precision thermometer
Thermometers Analogue Thermometers
This thermometer is suitable for government verification.
  • Solid stem
  • Top round finish
  • White backed
Toluene filling.
Certifikace: Conforms to DIN 12785.