Anti-IQ motif containing B1 Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM
A101013-100
ANTIA101013-100EA 475 EUR
ANTIA101013-100
Anti-IQ motif containing B1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to IQCB1 for WB and ELISA with samples derived from human.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: IQ motif containing B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR