Anti-Milk Fat Globule Mouse Monoclonal Antibody [clone: MFG-06]

Supplier: ANTIBODIES.COM
A249315-100
ANTIA249315-100EA 520 EUR
ANTIA249315-100
Anti-Milk Fat Globule Mouse Monoclonal Antibody [clone: MFG-06]
Antibodies
Mouse monoclonal [MFG-06] antibody to Milk Fat Globule for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Milk Fat Globule
Clonality: Monoclonal
Clone: MFG-06
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR