Anti-P Glycoprotein Rabbit Recombinant Antibody [clone: MDR1/8962R]

Supplier: ANTIBODIES.COM
A315431-100
ANTIA315431-100EA 680 EUR
ANTIA315431-100
Anti-P Glycoprotein Rabbit Recombinant Antibody [clone: MDR1/8962R]
Antibodies
Recombinant rabbit monoclonal [MDR1/8962R] antibody to P glycoprotein for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ml

Type: Primary
Antigen: P Glycoprotein
Clonality: Recombinant
Clone: MDR1/8962R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR