Indole-3-butyric acid potassium salt

Supplier: Apollo Scientific
Warning

Synonyms: Potassium 4-(1H-indol-3-yl)butanoate

BI5912-25G BI5912-10G BI5912-100G
APOSBI5912-25GEA 31.9 EUR
APOSBI5912-25G APOSBI5912-10G APOSBI5912-100G
Indole-3-butyric acid potassium salt
Indole-3-butyric acid potassium salt

  • Assay (by titration) 99+%
  • Water (by Karl Fischer) < 1.0%
  • Water soluble
  • Plant tissue culture tested

Formula: C₁₂H₁₂KNO₂
MW: 241.331 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00058442
CAS Number: 60096-23-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR