8-Aminopyrene-1,3,6-trisulphonic acid trisodium salt

Supplier: Apollo Scientific
Warning

Synonyms: 1,3,6-Pyrenetrisulphonic acid, 8-amino trisodium salt, Trisodium 8-aminopyrene-1,3,6-trisulphonate, APTS, 8-Aminopyrene-1,3,6-trisulphonic acid trisodium

BIB6285-500MG BIB6285-100MG BIB6285-250MG
APOSBIB6285-500MGEA 1150 EUR
APOSBIB6285-500MG APOSBIB6285-100MG APOSBIB6285-250MG
8-Aminopyrene-1,3,6-trisulphonic acid trisodium salt
8-Aminopyrene-1,3,6-trisulphonic acid trisodium salt
Formula: C₁₆H₈NNa₃O₉S₃
MW: 523.40 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00674494
CAS Number: 196504-57-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR