Phomalactone

Supplier: Apollo Scientific
Warning

BIBR1076-1MG
APOSBIBR1076-1MGEA 277 EUR
APOSBIBR1076-1MG
Phomalactone
Phomalactone
Formula: C₈H₁₀O₃
MW: 154.163 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02752490
CAS Number: 28921-94-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR