Cysteaminium chloride 99%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Aminoethanethiol hydrochloride, Cysteamine Hydrochloride, 2-Mercaptoethylamine hydrochloride, 2-MEA, β-Mercaptoethylamine hydrochloride, Mercaptoethylamine-HCl, H-Cys-NH2·HCl

BIC2510-100G BIC2510-25G
APOSBIC2510-100GEA 60.9 EUR
APOSBIC2510-100G APOSBIC2510-25G
Cysteaminium chloride 99%
Cysteaminium chloride
Formula: C₂H₇NS·HCl
MW: 113.61 g/mol
Melting Pt: 66…68 °C
Density: 0.75 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012904
CAS Number: 156-57-0
EINECS: 205-858-1
UN: 3335
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR