1-Azido-1-deoxy-β-D-glucopyranoside 98

Supplier: Apollo Scientific
Warning

Synonyms: β-D-Glucopyranosyl azide

BICL4025-1G BICL4025-500MG BICL4025-100MG
APOSBICL4025-1GEA 944 EUR
APOSBICL4025-1G APOSBICL4025-500MG APOSBICL4025-100MG
1-Azido-1-deoxy-β-D-glucopyranoside 98
1-Azido-1-deoxy-β-D-glucopyranoside
Formula: C₆H₁₁N₃O₅
MW: 205.169 g/mol
Melting Pt: 150 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD03701503
CAS Number: 20379-59-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR