β-D-Xylopyranosyl azide 98

Supplier: Apollo Scientific

BICL4245-250MG BICL4245-500MG BICL4245-100MG
APOSBICL4245-250MGEA 517 EUR
APOSBICL4245-250MG APOSBICL4245-500MG APOSBICL4245-100MG
β-D-Xylopyranosyl azide 98
β-D-Xylopyranosyl azide
Formula: C₅H₉N₃O₄
MW: 175.143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16621028
CAS Number: 51368-20-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR