α-D-Xylopyranosyl azide 98

Supplier: Apollo Scientific

BICL4270-100MG BICL4270-25MG BICL4270-50MG
APOSBICL4270-100MGEA 1270 EUR
APOSBICL4270-100MG APOSBICL4270-25MG APOSBICL4270-50MG
α-D-Xylopyranosyl azide 98
α-D-Xylopyranosyl azide
Formula: C₅H₉N₃O₄
MW: 175.143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28122827
CAS Number: 100842-21-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR