Vancomycin hydrochloride 100 mg/ml

Supplier: Apollo Scientific
Warning

Synonyms: (3S,6R,7R,22R,23S,26S,36R,38aR)-3-(2-Amino-2-oxoethyl)-44-[[2-O-(3-amino-2,3,6-trideoxy-3-C-methyl-α-L-lyxo-hexopyranosyl)-β-D-glucopyranosyl]oxy]-10,19-dichloro-2,3,4,5,6,7,23,24,25,26,36,37,38,38a-tetradecahydro-7,22,28,30,32-pentahydroxy-6-[[(2R)-4-methyl-2-(methylamino)-1-oxopentyl]amino]-2,5,24,38,39-pentaoxo-22H-8,11:18,21-Dietheno-23,36-(iminomethano)-13,16:31,35-dimetheno-1H,16H-[1,6,9]oxadiazacyclohexadecino[4,5-m][10,2,16]benzoxadiazacyclotetracosine-26-carboxylic acid xhydrochloride, Vancomycin supplement

BIG1035-10ML BIG1035-25ML BIG1035-100ML
APOSBIG1035-10MLEA 169 EUR
APOSBIG1035-10ML APOSBIG1035-25ML APOSBIG1035-100ML
Vancomycin hydrochloride 100 mg/ml
Vancomycin hydrochloride

  • Antibodies

Formula: C₆₆H₇₅Cl₂N₉O₂₄·HCl
MW: 1485.71 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03613611
CAS Number: 1404-93-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR