Methyl 6-amino-6-deoxy-a-D-glucopyranoside

Supplier: Apollo Scientific
Warning

BIM6003-100MG BIM6003-250MG BIM6003-500MG
APOSBIM6003-100MGEA 145 EUR
APOSBIM6003-100MG APOSBIM6003-250MG APOSBIM6003-500MG
Methyl 6-amino-6-deoxy-a-D-glucopyranoside
Methyl 6-amino-6-deoxy-a-D-glucopyranoside
Formula: C₇H₁₅NO₅
MW: 193.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04038151
CAS Number: 5155-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR