L-779450

Supplier: Apollo Scientific

Synonyms: 2-Chloro-5-(2-phenyl-5-(pyridin-4-yl)-1H-imidazol-4-yl)phenol, Raf Kinase Inhibitor IV, L 779450, 2-Chloro-5-(2-phenyl-4-(pyridin-4-yl)-1H-imidazol-5-yl)phenol, 2-(Phenyl)-4-(3-hydroxy-4-chlorophenyl)-5-(4-pyridyl)-1H-imidazole, L779450

BISN0240-50MG
APOSBISN0240-50MGEA 119 EUR
APOSBISN0240-50MG
L-779450
L-779450

MAPK/ERK Signalling Pathway

Formula: C₂₀H₁₄ClN₃O
MW: 347.80 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12828750
CAS Number: 303727-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR