TAPS (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid) 99

Supplier: Apollo Scientific
Warning

Synonyms: 3-{[2-Hydroxy-1,1-bis(hydroxymethyl)ethyl]amino}-1-propanesulphonic acid, N-Tris(hydroxymethyl)methyl-3-aminopropanesulphonic acid

BIT3005-100G BIT3005-1KG BIT3005-250G
APOSBIT3005-100GEA 29 EUR
APOSBIT3005-100G APOSBIT3005-1KG APOSBIT3005-250G
TAPS (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid) 99
TAPS (3-(Tris(hydroxymethyl)methylamino)propane-1-sulphonic acid)

Biological buffer - useful range 7.7-9.1


  • Assay (by titration) >99%
  • Water (by Karl Fischer) <1.0%

Formula: C₇H₁₇NO₆S
MW: 243.28 g/mol
Melting Pt: 230…235 °C (dec.)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007538
CAS Number: 29915-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR