Aluminium 99.999%

Supplier: Apollo Scientific
Warning

Synonyms: Aluminum

IN1003-1KG IN1003-250G
APOSIN1003-1KGEA 0 EUR
APOSIN1003-1KG APOSIN1003-250G
Aluminium 99.999%
Aluminium
Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2467 °C
Melting Pt: 660.1 °C
Density: 2.699 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
UN: 3178
ADR: 4.1 III
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR