Aluminium oxide 99.999%

Supplier: Apollo Scientific
Warning

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

IN1039-100G
APOSIN1039-100GEA 625 EUR
APOSIN1039-100G
Aluminium oxide 99.999%
Aluminium oxide
Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980 °C
Melting Pt: 2072 °C
Density: 3.97 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
Merck Index: 13,00355

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR