Lithium hydroxide monohydrate 98%

Supplier: Apollo Scientific
Danger

IN2340-2.5KG IN2340-500G
APOSIN2340-2.5KGEA 1210 EUR
APOSIN2340-2.5KG APOSIN2340-500G
Lithium hydroxide monohydrate 98%
Lithium hydroxide monohydrate
Formula: LiOH·1H₂O
MW: 41.96 g/mol
Boiling Pt: 924 °C
Melting Pt: 462 °C
Density: 1.51 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149772
CAS Number: 1310-66-3
EINECS: 215-183-4
UN: 2680
ADR: 8 II
Merck Index: 13,05556

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR