5-(Fur-2-yl)-1-phenyl-1H-pyrazole-3-carboxylic acid

Supplier: Apollo Scientific

OR01493-500MG
APOSOR01493-500MGEA 313 EUR
APOSOR01493-500MG
5-(Fur-2-yl)-1-phenyl-1H-pyrazole-3-carboxylic acid
5-(Fur-2-yl)-1-phenyl-1H-pyrazole-3-carboxylic acid
Formula: C₁₄H₁₀N₂O₃
MW: 254.241 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06245333
CAS Number: 100537-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR