Phenyl(phenylamino)acetonitrile

Supplier: Apollo Scientific
Warning

Synonyms: Anilino(phenyl)acetonitrile

OR01615-100MG
APOSOR01615-100MGEA 65.4 EUR
APOSOR01615-100MG
Phenyl(phenylamino)acetonitrile
Phenyl(phenylamino)acetonitrile
Formula: C₁₄H₁₂N₂
MW: 208.258 g/mol
Melting Pt: 74…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00019793
CAS Number: 4553-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR