N-Fmoc-L-leucine

Supplier: Apollo Scientific
Warning

OR0491-500G
APOSOR0491-500GEA 192 EUR
APOSOR0491-500G
N-Fmoc-L-leucine
N-Fmoc-L-leucine
Formula: C₂₁H₂₃NO₄
MW: 353.4 g/mol
Melting Pt: 152…156 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037133
CAS Number: 35661-60-0
EINECS: 252-662-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR