3-Chloropropylamine hydrochloride 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Chloropropan-1-amine hydrochloride

OR10945-100G OR10945-500G
APOSOR10945-100GEA 21.9 EUR
APOSOR10945-100G APOSOR10945-500G
3-Chloropropylamine hydrochloride 98%
3-Chloropropylamine hydrochloride
Formula: C₃H₈ClN
MW: 130.01 g/mol
Melting Pt: 145…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00012913
CAS Number: 6276-54-6
EINECS: 228-474-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR