5-Bromo-3-chloropyrazin-2-amine

Supplier: Apollo Scientific

OR110341-25G
APOSOR110341-25GEA 63.8 EUR
APOSOR110341-25G
5-Bromo-3-chloropyrazin-2-amine
5-Bromo-3-chloropyrazin-2-amine
Formula: C₄H₃BrClN₃
MW: 208.4500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11109828
CAS Number: 21943-13-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR