6-Bromo-2-tert-butylimidazo[1,2-a]pyridine

Supplier: Apollo Scientific

OR110922-1G
APOSOR110922-1GEA 248 EUR
APOSOR110922-1G
6-Bromo-2-tert-butylimidazo[1,2-a]pyridine
6-Bromo-2-tert-butylimidazo[1,2-a]pyridine
Formula: C₁₁H₁₃BrN₂
MW: 253.138 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11036007
CAS Number: 864867-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR