7-Methylimidazo[1,2-a]pyridine-8-carbonitrile hydrochloride

Supplier: Apollo Scientific
Warning

OR111210-1G
APOSOR111210-1GEA 568 EUR
APOSOR111210-1G
7-Methylimidazo[1,2-a]pyridine-8-carbonitrile hydrochloride
7-Methylimidazo[1,2-a]pyridine-8-carbonitrile hydrochloride
Formula: C₉H₇N₃·ClH
MW: 193.633 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29918589
CAS Number: 2108781-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR