Indole-2-carboxylic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 1H-Indole-2-carboxylic acid

OR11262-10G
APOSOR11262-10GEA 21.9 EUR
APOSOR11262-10G
Indole-2-carboxylic acid 98%
Indole-2-carboxylic acid
Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: 202…206 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005611
CAS Number: 1477-50-5
EINECS: 216-030-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR