6-Bromopicolinamide 98%

Supplier: Apollo Scientific
Warning

Synonyms: 6-Bromopyridine-2-carboxamide

OR11548-5G
APOSOR11548-5GEA 133 EUR
APOSOR11548-5G
6-Bromopicolinamide 98%
6-Bromopicolinamide
Formula: C₆H₅BrN₂O
MW: 201.02 g/mol
Melting Pt: 133…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00234154
CAS Number: 25194-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR