6,8-Dichloro-3-(4-nitrophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone

Supplier: Apollo Scientific

OR12075-1G
APOSOR12075-1GEA 180 EUR
APOSOR12075-1G
6,8-Dichloro-3-(4-nitrophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
6,8-Dichloro-3-(4-nitrophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Formula: C₁₄H₇Cl₂N₃O₃S
MW: 368.2 g/mol
Melting Pt: 282…284 °C
Storage Temperature: Ambient
MDL Number: MFCD08689701
CAS Number: 937601-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR