3-(Phenoxymethyl)benzeneboronic acid, pinacol ester

Supplier: Apollo Scientific
Warning

Synonyms: 2-[3-(Phenoxymethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR12237-1G OR12237-250MG
APOSOR12237-1GEA 464 EUR
APOSOR12237-1G APOSOR12237-250MG
3-(Phenoxymethyl)benzeneboronic acid, pinacol ester
3-(Phenoxymethyl)benzeneboronic acid, pinacol ester
Formula: C₁₉H₂₃BO₃
MW: 310.195 g/mol
Melting Pt: 76.5…77.5 °C
Storage Temperature: Ambient
MDL Number: MFCD09065022
CAS Number: 912569-68-7
UN: 2811
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR