2-Phenylmorpholine hydrochloride

Supplier: Apollo Scientific
Warning

Synonyms: (Morpholin-2-yl)benzene hydrochloride

OR12786-1G OR12786-5G OR12786-10G
APOSOR12786-1GEA 143 EUR
APOSOR12786-1G APOSOR12786-5G APOSOR12786-10G
2-Phenylmorpholine hydrochloride
2-Phenylmorpholine hydrochloride
Formula: C₁₀H₁₃NO·ClH
MW: 199.677 g/mol
Melting Pt: 148…151 °C
Storage Temperature: Ambient
MDL Number: MFCD03869024
CAS Number: 23972-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR