3-(Chloromethyl)-5-methylisoxazole

Supplier: Apollo Scientific
Danger

OR1308-1G OR1308-5G OR1308-250MG
APOSOR1308-1GEA 69.6 EUR
APOSOR1308-1G APOSOR1308-5G APOSOR1308-250MG
3-(Chloromethyl)-5-methylisoxazole
3-(Chloromethyl)-5-methylisoxazole
Formula: C₅H₆ClNO
MW: 131.560 g/mol
Boiling Pt: 93 °C/15mmHg
Storage Temperature: Ambient
MDL Number: MFCD00221166
CAS Number: 35166-37-1
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR